Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RHCE SIRNA Assays ›
          Gene Symbol:
          RHCE RHD
          Gene Name:
          Rh blood group CcEe antigens
          Rh blood group D antigen
          Gene Aliases:
          CD240CE, RH, RH30A, RHC, RHE, RHIXB, RHPI, Rh4, RhIVb(J), RhVI, RhVIII
          CD240D, DIIIc, RH, RH30, RHCED, RHDVA(TT), RHDel, RHPII, RHXIII, Rh4, RhDCw, RhII, RhK562-II, RhPI
          Chromosome Location:
          Chr.1: 25362249 - 25430193 on Build GRCh38
          Chromosome Location:
          Chr.1: 25272393 - 25330445 on Build GRCh38
          UniGene ID:
          Hs.449968
          Hs.449968
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS143637
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RHCE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020485.4 NP_065231.3 3 464
          NM_138616.3 NP_619522.3 3 464
          NM_138617.3 NP_619523.3 3 464
          NM_138618.3 NP_619524.3 3 464
          XM_005245957.3 XP_005246014.1 3 929
          XM_006710810.3 XP_006710873.1 3 933
          XM_011541888.2 XP_011540190.1 4 964
          XM_011541889.2 XP_011540191.1 3 479
          XM_011541890.2 XP_011540192.1 3 925
          XM_011541891.2 XP_011540193.1 4 581
          XM_017002014.1 XP_016857503.1 3 932
          GenBank mRNA AB018644.1 - 3 378
          AB018645.1 - 3 378
          AF056965.1 - 3 378
          AF510065.1 - 3 421
          AF510066.1 - 3 421
          AF510067.1 - 3 421
          AF510068.1 - 3 421
          AM295498.2 - 3 378
          AM295506.2 - 3 378
          AY603478.1 - 3 394
          BC075081.2 - 3 399 30915384
          BC139905.1 - 3 506 40147407
          DQ178642.1 - 3 378
          DQ266353.1 - 3 378
          DQ266400.1 - 3 378
          DQ322275.1 - 3 378
          FR691750.1 - 3 378
          GU474431.1 - 3 378
          GU563377.1 - 3 378
          GU810838.1 - 3 378
          HE572747.1 - 3 378
          HG326207.1 - 3 378
          HQ711843.1 - 3 378
          HQ711844.1 - 3 398
          HQ711845.1 - 3 378
          HQ711846.1 - 3 378
          HQ711847.1 - 3 398
          JX420291.1 - 3 378
          JX420292.1 - 3 378
          JX420293.1 - 3 378
          JX420294.1 - 3 378
          JX420295.1 - 3 378
          JX420296.1 - 3 378
          JX420298.1 - 3 378
          JX420299.1 - 3 378
          JX420300.1 - 3 378
          JX420301.1 - 3 378
          JX420302.1 - 3 378
          KM975480.1 - 3 378
          M34015.1 - 3 408
          S57967.1 - 3 378
          X54534.1 - 3 422
          X63095.1 - 3 387
          X63096.1 - 3 384
          X63098.1 - 3 397
          Gene Symbol: RHD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB049754.1 - 3 378
          AF312679.1 - 3 413
          S82449.1 - 3 378
          Z97026.1 - 3 398

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          organic cation transport
          nitrogen utilization
          ammonium transmembrane transport
          ammonium transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0