Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SIRT6 SIRNA Assays ›
          Gene Symbol:
          SIRT6
          Gene Name:
          sirtuin 6
          Gene Aliases:
          SIR2L6
          Chromosome Location:
          Chr.19: 4174109 - 4182604 on Build GRCh38
          UniGene ID:
          Hs.423756
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS147498
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SIRT6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193285.2 NP_001180214.1 6 705
          NM_001321058.1 NP_001307987.1 6 658
          NM_001321059.1 NP_001307988.1 6 603
          NM_001321060.1 NP_001307989.1 7 746
          NM_001321061.1 NP_001307990.1 5 577
          NM_001321062.1 NP_001307991.1 5 475
          NM_001321063.1 NP_001307992.1 6 563
          NM_001321064.1 NP_001307993.1 7 711
          NM_016539.3 NP_057623.2 7 786
          GenBank mRNA AF233396.1 - 7 781
          AK074810.1 - 7 749
          AK225567.1 - 7 754
          AK293348.1 - 6 619
          AK299420.1 - 5 1129
          AK301247.1 - 4 1175
          AK315048.1 - 7 750
          AL365375.1 - 185
          AL559383.3 - 7 685
          BC004218.2 - 5 517 3352203
          BC005026.1 - 7 724 2820472
          BC028220.1 - 6 651 5217599
          BE281103.1 - 6 544
          BP418952.1 - 5 420
          BQ060703.1 - 6 499
          CN364175.1 - 7 689
          CR457200.1 - 7 721

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          post-embryonic cardiac muscle cell growth involved in heart morphogenesis
          base-excision repair
          protein ADP-ribosylation
          negative regulation of cell proliferation
          regulation of double-strand break repair via homologous recombination
          protein destabilization
          response to nutrient levels
          glucose homeostasis
          negative regulation of glycolytic process
          negative regulation of transcription, DNA-templated
          negative regulation of glucose import
          positive regulation of fibroblast proliferation
          histone H3-K9 modification
          histone H3-K9 deacetylation
          positive regulation of stem cell proliferation
          chromatin binding
          transcription corepressor activity
          NAD+ ADP-ribosyltransferase activity
          NAD(P)+-protein-arginine ADP-ribosyltransferase activity
          protein binding
          zinc ion binding
          NAD-dependent histone deacetylase activity
          NAD-dependent protein deacetylase activity
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          NAD+ binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0