Search
Search

LYPLA2| Gene Symbol: LYPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007260.2 | NP_009191.1 | 2, 3 | 192 | ||
| XM_005245728.4 | XP_005245785.1 | 2, 3 | 664 | |||
| XM_017000165.1 | XP_016855654.1 | 2, 3 | 559 | |||
| GenBank mRNA | AF098668.1 | - | 2, 3 | 192 | ||
| BC017034.2 | - | 2, 3 | 157 | 3845764 | ||
| BC017193.2 | - | 2, 3 | 146 | 3138786 | ||