Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009880.1 | NP_001009880.1 | 2 | 170 | ||
NM_015264.1 | NP_056079.1 | 2 | 242 | |||
XM_011530017.2 | XP_011528319.1 | 2 | 205 | |||
XM_017028672.1 | XP_016884161.1 | 2 | 214 | |||
GenBank mRNA | AI830875.1 | - | 2 | 238 | ||
AK094937.1 | - | 3 | 253 | |||
AK124648.1 | - | 2 | 250 | |||
AK297058.1 | - | 2 | 222 | |||
AK309015.1 | - | 1 | 294 | |||
AK315883.1 | - | 2 | 154 | |||
BC038414.1 | - | 1 | 317 | |||
BC053595.1 | - | 2 | 164 | 5730020 | ||
CR456402.1 | - | 2 | 107 | |||
KC154032.1 | - | 2 | 170 |