Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADM1 SIRNA Assays ›
Gene Symbol:
CADM1
Gene Name:
cell adhesion molecule 1
Gene Aliases:
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
Chromosome Location:
Chr.11: 115173625 - 115504523 on Build GRCh38
UniGene ID:
Hs.370510
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177559
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098517.1
NP_001091987.1
7
1107
NM_001301043.1
NP_001287972.1
7
1107
NM_001301044.1
NP_001287973.1
7
1107
NM_001301045.1
NP_001287974.1
7
1107
NM_014333.3
NP_055148.3
7
1107
XM_005271494.2
XP_005271551.1
7
1000
XM_017017457.1
XP_016872946.1
7
1066
XM_017017458.1
XP_016872947.1
7
873
XM_017017459.1
XP_016872948.1
8
3060
XM_017017460.1
XP_016872949.1
8
3060
XM_017017461.1
XP_016872950.1
8
1074
GenBank mRNA
AB094146.1
-
7
978
AF132811.1
-
7
981
AK075502.1
-
7
997
AK315245.1
-
7
1040
AY309502.1
-
2
175
AY358334.1
-
7
993
BC035930.1
-
7
999
BC047021.1
-
7
1107
5248100
BC125102.1
-
7
982
BC125103.1
-
7
981
40118330
CR457157.1
-
7
978
HE586496.1
-
7
981
HE586497.1
-
7
981
HE586498.1
-
7
981
HE586499.1
-
7
981
HE586500.1
-
8
1067
HE586501.1
-
1
98
HE586503.1
-
8
1009
KJ534791.1
-
7
978
KJ534792.1
-
7
978
KJ534793.1
-
7
978
KJ534794.1
-
7
978
KJ534795.1
-
7
978
KJ534796.1
-
7
978
KJ534797.1
-
7
978
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
T cell mediated cytotoxicity
apoptotic process
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
multicellular organism development
spermatogenesis
cell recognition
cell differentiation
adherens junction organization
susceptibility to natural killer cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of cytokine secretion
activated T cell proliferation
detection of stimulus
receptor activity
receptor binding
protein binding
PDZ domain binding
protein homodimerization activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0