Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135197.1 | NP_001128669.1 | 8 | 1121 | ||
NM_178173.3 | NP_835467.2 | 10 | 1419 | |||
XM_006713124.3 | XP_006713187.1 | 8 | 1296 | |||
XM_011533670.2 | XP_011531972.1 | 7 | 1130 | |||
XM_011533671.2 | XP_011531973.1 | 7 | 1124 | |||
XM_011533672.2 | XP_011531974.1 | 8 | 1489 | |||
XM_011533673.2 | XP_011531975.1 | 8 | 1521 | |||
GenBank mRNA | AK058049.1 | - | 10 | 1310 | ||
AK303033.1 | - | 8 | 1121 | |||
BC036202.1 | - | 10 | 1419 | 4836976 |