Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MSX2 SIRNA Assays ›
          Gene Symbol:
          MSX2
          Gene Name:
          msh homeobox 2
          Gene Aliases:
          CRS2, FPP, HOX8, MSH, PFM, PFM1
          Chromosome Location:
          Chr.5: 174724495 - 174730899 on Build GRCh38
          UniGene ID:
          Hs.89404
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181360
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MSX2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002449.4 NP_002440.2 1 396
          XM_017009489.1 XP_016864978.1 1 473
          GenBank mRNA AL552909.3 - 1 397
          BC015509.1 - 1 331 3903028
          BT009814.1 - 1 308
          D14970.1 - 1 23
          D26145.1 - 1 331
          D31771.1 - 1 369
          D89377.1 - 1 369
          S75361.1 - 1 369

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012982, BF414483.
          This siRNA also targets the following transcripts in Mouse: NM_013601, X59252, BC141132, AK131974, AK133846, and others.

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          osteoblast differentiation
          chondrocyte development
          osteoblast development
          outflow tract septum morphogenesis
          epithelial to mesenchymal transition involved in endocardial cushion formation
          endochondral bone growth
          transcription from RNA polymerase II promoter
          negative regulation of cell proliferation
          anterior/posterior pattern specification
          signal transduction involved in regulation of gene expression
          positive regulation of BMP signaling pathway
          negative regulation of CREB transcription factor activity
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          wound healing, spreading of epidermal cells
          embryonic nail plate morphogenesis
          negative regulation of apoptotic process
          negative regulation of fat cell differentiation
          negative regulation of keratinocyte differentiation
          positive regulation of osteoblast differentiation
          negative regulation of transcription, DNA-templated
          stem cell differentiation
          positive regulation of catagen
          bone trabecula formation
          cranial suture morphogenesis
          frontal suture morphogenesis
          branching involved in mammary gland duct morphogenesis
          BMP signaling pathway involved in heart development
          enamel mineralization
          cellular response to estradiol stimulus
          activation of meiosis
          negative regulation of transcription regulatory region DNA binding
          positive regulation of mesenchymal cell apoptotic process
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcription cofactor activity
          protein binding
          transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0