Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STAT6 SIRNA Assays ›
          Gene Symbol:
          STAT6
          Gene Name:
          signal transducer and activator of transcription 6
          Gene Aliases:
          D12S1644, IL-4-STAT, STAT6B, STAT6C
          Chromosome Location:
          Chr.12: 57095404 - 57111433 on Build GRCh38
          UniGene ID:
          Hs.524518
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186136
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STAT6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178078.1 NP_001171549.1 18 2372
          NM_001178079.1 NP_001171550.1 18 2298
          NM_001178080.1 NP_001171551.1 17 2216
          NM_001178081.1 NP_001171552.1 16 2077
          NM_003153.4 NP_003144.3 18 2353
          NR_033659.1 17 2214
          XM_006719575.2 XP_006719638.1 19 2350
          XM_011538703.2 XP_011537005.1 19 2427
          XM_011538704.2 XP_011537006.1 20 2438
          XM_011538705.2 XP_011537007.1 20 2548
          XM_011538706.2 XP_011537008.1 20 2404
          XM_011538707.2 XP_011537009.1 19 2389
          XM_011538708.2 XP_011537010.1 18 2231
          XM_011538709.2 XP_011537011.1 19 2265
          GenBank mRNA AB103089.1 - 12 1525
          AK290431.1 - 18 2301
          AK291044.1 - 18 2299
          AK298721.1 - 16 1897
          AK299094.1 - 17 2150
          AK308751.1 - 18 2193
          AK312471.1 - 18 2219
          AK316142.1 - 17 2160
          AK316236.1 - 17 1991
          AY615283.1 - 4 931
          BC075852.1 - 18 2154 5530399
          U16031.1 - 18 2212

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          T-helper 1 cell lineage commitment
          negative regulation of type 2 immune response
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          signal transduction
          positive regulation of type I interferon production
          mammary gland epithelial cell proliferation
          interleukin-4-mediated signaling pathway
          regulation of cell proliferation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of isotype switching to IgE isotypes
          mammary gland morphogenesis
          cellular response to hydrogen peroxide
          cellular response to reactive nitrogen species
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          protein binding
          protein phosphatase binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-n24h2:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0