Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MSMP SIRNA Assays ›
          Gene Symbol:
          MSMP NPR2 RGP1
          Gene Name:
          microseminoprotein, prostate associated
          natriuretic peptide receptor 2
          RGP1 homolog, RAB6A GEF complex partner 1
          Gene Aliases:
          PSMP
          AMDM, ANPRB, ANPb, ECDM, GUC2B, GUCY2B, NPRB, NPRBi, SNSK
          KIAA0258
          Chromosome Location:
          Chr.9: 35752990 - 35754277 on Build GRCh38
          Chromosome Location:
          Chr.9: 35752945 - 35809731 on Build GRCh38
          Chromosome Location:
          Chr.9: 35749280 - 35753267 on Build GRCh38
          UniGene ID:
          Hs.571601
          Hs.78518
          Hs.493804
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID HSS186224
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MSMP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001044264.2 NP_001037729.1 1 164
          GenBank mRNA BC105274.1 - 1 168
          DQ012170.1 - 1 162
          Gene Symbol: NPR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005251479.4 XP_005251536.1 1 1166
          XM_011517890.2 XP_011516192.1 1 1166
          XM_011517891.2 XP_011516193.1 1 1166
          XM_011517892.2 XP_011516194.1 1 1166
          XM_011517893.2 XP_011516195.1 1 1166
          XM_017014745.1 XP_016870234.1 1 1166
          XM_017014746.1 XP_016870235.1 1 1166
          XM_017014747.1 XP_016870236.1 1 1166
          XM_017014748.1 XP_016870237.1 1 1166
          XM_017014749.1 XP_016870238.1 1 1166
          XM_017014750.1 XP_016870239.1 1 1166
          Gene Symbol: RGP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA D87447.1 - 9 2497

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule guanylate cyclase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          ossification
          cGMP biosynthetic process
          protein phosphorylation
          receptor guanylyl cyclase signaling pathway
          regulation of blood pressure
          intracellular signal transduction
          single organism reproductive process
          negative regulation of meiotic cell cycle
          bone development
          cellular response to granulocyte macrophage colony-stimulating factor stimulus
          negative regulation of oocyte maturation
          regulation of cardiac conduction
          molecular_function
          adenylate cyclase activity
          guanylate cyclase activity
          protein kinase activity
          ATP binding
          GTP binding
          natriuretic peptide receptor activity
          peptide hormone binding
          hormone binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0