Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCOF1 SIRNA Assays ›
          Gene Symbol:
          TCOF1
          Gene Name:
          treacle ribosome biogenesis factor 1
          Gene Aliases:
          MFD1, TCS, TCS1, treacle
          Chromosome Location:
          Chr.5: 150357639 - 150400308 on Build GRCh38
          UniGene ID:
          Hs.519672
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186249
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCOF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000356.3 NP_000347.2 5 551
          NM_001008657.2 NP_001008657.1 5 551
          NM_001135243.1 NP_001128715.1 5 551
          NM_001135244.1 NP_001128716.1 5 551
          NM_001135245.1 NP_001128717.1 5 551
          NM_001195141.1 NP_001182070.1 5 551
          XM_005268502.3 XP_005268559.1 5 488
          XM_005268503.3 XP_005268560.1 5 488
          XM_005268504.3 XP_005268561.1 5 488
          XM_005268505.3 XP_005268562.1 5 488
          XM_005268506.3 XP_005268563.1 5 488
          XM_005268507.3 XP_005268564.1 5 488
          XM_005268508.3 XP_005268565.1 5 488
          XM_005268509.3 XP_005268566.1 5 488
          XM_011537678.2 XP_011535980.1 5 488
          XM_017009792.1 XP_016865281.1 5 488
          XM_017009793.1 XP_016865282.1 5 488
          XM_017009794.1 XP_016865283.1 5 488
          XM_017009795.1 XP_016865284.1 5 488
          GenBank mRNA AK096467.1 - 1 57
          AK225284.1 - 5 476
          AK303611.1 - 5 476
          AK307586.1 - 5 476
          AY460334.1 - 5 443
          BC011764.2 - 5 472 3616898
          BC014559.2 - 5 472 4025673
          BC016144.1 - 5 471
          BC027252.1 - 5 399
          BC033093.1 - 5 472 4577077
          BX537490.1 - 1 1208
          U40847.1 - 5 536
          U76366.1 - 5 518

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          transport
          transporter activity
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0