Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTMR3 SIRNA Assays ›
          Gene Symbol:
          MTMR3
          Gene Name:
          myotubularin related protein 3
          Gene Aliases:
          FYVE-DSP1, ZFYVE10
          Chromosome Location:
          Chr.22: 29883165 - 30030868 on Build GRCh38
          UniGene ID:
          Hs.474536
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS189646
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MTMR3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021090.3 NP_066576.1 20 3834
          NM_153050.2 NP_694690.1 20 3750
          NM_153051.2 NP_694691.1 19 3723
          XM_005261803.2 XP_005261860.1 22 3979
          XM_005261804.3 XP_005261861.1 21 3896
          XM_005261805.2 XP_005261862.1 21 3952
          XM_005261806.2 XP_005261863.1 21 3861
          XM_005261808.2 XP_005261865.1 20 3841
          XM_017029028.1 XP_016884517.1 20 3753
          XM_017029029.1 XP_016884518.1 19 3716
          XM_017029030.1 XP_016884519.1 19 3648
          XM_017029031.1 XP_016884520.1 18 3609
          XM_017029032.1 XP_016884521.1 19 3606
          XM_017029033.1 XP_016884522.1 15 3180
          XM_017029034.1 XP_016884523.1 21 5183
          GenBank mRNA AB002369.1 - 20 3753
          AF233436.2 - 17 3431
          AF233437.1 - 16 3404
          AF233438.1 - 17 3515
          AK289581.1 - 20 3834
          BC142713.1 - 19 3679 40148839
          BC148216.1 - 18 3542 40082266
          BC151217.1 - 20 3848 40148098
          BC152455.1 - 20 3753 8862588
          CR456525.1 - 18 3530

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          phosphatidylinositol biosynthetic process
          macroautophagy
          peptidyl-tyrosine dephosphorylation
          cellular response to glucose starvation
          phosphatidylinositol dephosphorylation
          phosphatidylinositol 5-phosphate metabolic process
          regulation of autophagosome assembly
          phosphatidylinositol-3-phosphatase activity
          protein serine/threonine phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          metal ion binding
          phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0