Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KRIT1 SIRNA Assays ›
Gene Symbol:
KRIT1
Gene Name:
KRIT1, ankyrin repeat containing
Gene Aliases:
CAM, CCM1
Chromosome Location:
Chr.7: 92198969 - 92246128 on Build GRCh38
UniGene ID:
Hs.531987
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189649
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KRIT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013406.1
NP_001013424.1
15
2087
NM_004912.3
NP_004903.2
17
2726
NM_194454.1
NP_919436.1
17
2379
NM_194455.1
NP_919437.1
17
2853
NM_194456.1
NP_919438.1
18
2618
XM_005250660.3
XP_005250717.1
17
2346
XM_005250662.3
XP_005250719.1
17
2325
XM_005250665.3
XP_005250722.1
17
2253
XM_005250666.3
XP_005250723.1
18
2429
XM_005250667.2
XP_005250724.1
16
2137
XM_005250668.3
XP_005250725.1
16
2199
XM_005250669.3
XP_005250726.1
16
2177
XM_006716161.3
XP_006716224.1
17
2477
XM_006716162.3
XP_006716225.1
17
2294
XM_006716163.3
XP_006716226.1
16
2144
XM_011516651.2
XP_011514953.1
18
2460
XM_011516653.2
XP_011514955.1
16
2214
XM_011516654.2
XP_011514956.1
18
2478
XM_011516655.2
XP_011514957.1
16
2132
XM_011516656.2
XP_011514958.1
16
2101
XM_011516657.2
XP_011514959.1
16
2021
XM_011516658.2
XP_011514960.1
16
2033
XM_011516659.2
XP_011514961.1
16
2055
XM_011516660.2
XP_011514962.1
15
1937
XM_011516661.2
XP_011514963.1
15
2015
XM_017012755.1
XP_016868244.1
16
2104
XM_017012756.1
XP_016868245.1
16
2066
XM_017012757.1
XP_016868246.1
16
2098
XM_017012758.1
XP_016868247.1
17
2573
XM_017012759.1
XP_016868248.1
16
2425
GenBank mRNA
AF296765.2
-
17
2623
AF310133.1
-
17
2318
AF388384.1
-
17
2853
AK055305.1
-
15
2087
AK056537.1
-
16
2161
AK226131.1
-
10
1476
AY380057.1
-
17
2318
AY380058.1
-
17
2318
AY380059.1
-
17
2317
AY380060.1
-
17
2316
AY993944.1
-
14
1834
BC094684.1
-
14
1844
6174749
BX648659.1
-
11
2358
U90268.1
-
10
1238
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of endothelial cell proliferation
small GTPase mediated signal transduction
negative regulation of endothelial cell migration
negative regulation of angiogenesis
positive regulation of protein binding
cell redox homeostasis
regulation of catalytic activity
regulation of establishment of cell polarity
negative regulation of endothelial cell apoptotic process
protein binding
phosphatidylinositol-4,5-bisphosphate binding
microtubule binding
GTPase regulator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0