Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KRIT1 SIRNA Assays ›
Gene Symbol:
KRIT1
Gene Name:
KRIT1, ankyrin repeat containing
Gene Aliases:
CAM, CCM1
Chromosome Location:
Chr.7: 92198969 - 92246128 on Build GRCh38
UniGene ID:
Hs.531987
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189650
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KRIT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013406.1
NP_001013424.1
7
977
NM_004912.3
NP_004903.2
8
1472
NM_194454.1
NP_919436.1
8
1125
NM_194455.1
NP_919437.1
8
1599
NM_194456.1
NP_919438.1
9
1364
XM_005250660.3
XP_005250717.1
8
1092
XM_005250662.3
XP_005250719.1
8
1071
XM_005250665.3
XP_005250722.1
8
999
XM_005250666.3
XP_005250723.1
9
1175
XM_005250667.2
XP_005250724.1
7
883
XM_005250668.3
XP_005250725.1
7
945
XM_005250669.3
XP_005250726.1
7
923
XM_006716161.3
XP_006716224.1
8
1223
XM_006716162.3
XP_006716225.1
8
1040
XM_006716163.3
XP_006716226.1
7
890
XM_011516651.2
XP_011514953.1
9
1206
XM_011516653.2
XP_011514955.1
7
960
XM_011516654.2
XP_011514956.1
9
1224
XM_011516655.2
XP_011514957.1
7
878
XM_011516656.2
XP_011514958.1
7
847
XM_011516657.2
XP_011514959.1
7
767
XM_011516658.2
XP_011514960.1
7
779
XM_011516659.2
XP_011514961.1
7
801
XM_011516660.2
XP_011514962.1
6
683
XM_011516661.2
XP_011514963.1
6
761
XM_017012755.1
XP_016868244.1
7
850
XM_017012756.1
XP_016868245.1
7
812
XM_017012757.1
XP_016868246.1
7
844
XM_017012758.1
XP_016868247.1
8
1450
XM_017012759.1
XP_016868248.1
7
1302
GenBank mRNA
AF296765.2
-
8
1369
AF310133.1
-
8
1064
AF388384.1
-
8
1599
AJ294850.1
-
9
1323
AK055305.1
-
7
977
AK056537.1
-
7
907
AK226131.1
-
1
222
AY380057.1
-
8
1064
AY380058.1
-
8
1064
AY380059.1
-
8
1064
AY380060.1
-
8
1064
AY993944.1
-
5
580
BC094684.1
-
5
590
6174749
BX648659.1
-
3
1212
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of endothelial cell proliferation
small GTPase mediated signal transduction
negative regulation of endothelial cell migration
negative regulation of angiogenesis
positive regulation of protein binding
cell redox homeostasis
regulation of catalytic activity
regulation of establishment of cell polarity
negative regulation of endothelial cell apoptotic process
protein binding
phosphatidylinositol-4,5-bisphosphate binding
microtubule binding
GTPase regulator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0