Search
Search

Hpse| Gene Symbol: Hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152803.4 | NP_690016.1 | 2, 3 | 427 | ||
| GenBank mRNA | AF359507.1 | - | 2, 3 | 416 | ||
| AK040471.1 | - | 2, 3 | 428 | |||
| AK087283.1 | - | 2, 3 | 454 | |||
| AK154628.1 | - | 2, 3 | 428 | |||
| AY077467.1 | - | 2, 3 | 419 | |||
| AY151051.1 | - | 2, 3 | 338 | |||
| BC138782.1 | - | 2, 3 | 421 | 8861804 | ||
| BC138784.1 | - | 2, 3 | 421 | 8861806 | ||