Search
Search

Cog1| Gene Symbol: Cog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013581.3 | NP_038609.3 | 7 | 1352 | ||
| GenBank mRNA | AF109377.2 | - | 7 | 1335 | ||
| AK031510.1 | - | 6 | 808 | |||
| AK034636.2 | - | 7 | 1357 | |||
| AK152464.1 | - | 7 | 1343 | |||
| AK158507.1 | - | 7 | 1714 | |||
| AK159497.1 | - | 7 | 1354 | |||
| AK162971.1 | - | 7 | 1353 | |||
| AK171574.1 | - | 7 | 1346 | |||
| AK191517.1 | - | 2 | 95 | |||
| AK199318.1 | - | 2 | 113 | |||
| AK220387.1 | - | 7 | 1442 | |||
| BC063056.1 | - | 7 | 1334 | 5720460 | ||