Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141813.1 | - | 13 | 1662 | ||
AK170166.1 | - | 13 | 1988 | |||
AK172083.1 | - | 12 | 1421 | |||
BC063768.1 | - | 13 | 1642 | |||
BQ828670.1 | - | 1 | 29 | |||
U14166.1 | - | 13 | 1685 | |||
U15159.1 | - | 13 | 1657 |