Search
Search

Serpine2| Gene Symbol: Serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009255.4 | NP_033281.1 | 2 | 420 | ||
| GenBank mRNA | AK045954.1 | - | 2 | 392 | ||
| AK047856.1 | - | 2 | 392 | |||
| AK051332.1 | - | 2 | 392 | |||
| AK159687.1 | - | 2 | 392 | |||
| AK160700.1 | - | 2 | 392 | |||
| AK164270.1 | - | 2 | 392 | |||
| BC010675.1 | - | 2 | 370 | 3595407 | ||
| CT010311.1 | - | 1 | 243 | |||
| X70296.1 | - | 2 | 391 | |||