Search
Search

Cep104| Gene Symbol: Cep104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177673.2 | NP_808341.1 | 4, 5 | 509 | ||
| GenBank mRNA | AK129163.1 | - | 3, 4 | 375 | ||
| AK143802.1 | - | 4, 5 | 924 | |||
| AK149826.1 | - | 4, 5 | 860 | |||
| AK185711.1 | - | 1, 2 | 103 | |||
| BC046331.1 | - | 4, 5 | 509 | 6485859 | ||