Search
Search

Spata21| Gene Symbol: Spata21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177867.3 | NP_808535.1 | 8 | 1543 | ||
| GenBank mRNA | AK077155.1 | - | 8 | 1544 | ||
| BC112427.1 | - | 7 | 1252 | 40062127 | ||
| BC113132.1 | - | 7 | 1252 | 40062125 | ||