Search
Search

Tmem147| Gene Symbol: Tmem147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027215.2 | NP_081491.2 | 6 | 569 | ||
| GenBank mRNA | AK007753.1 | - | 6 | 530 | ||
| AK008089.1 | - | 6 | 523 | |||
| AK010168.1 | - | 6 | 558 | |||
| AK160267.1 | - | 6 | 573 | |||
| BC035323.1 | - | 6 | 475 | |||
| BC046908.1 | - | 2 | 75 | |||
| BC147300.1 | - | 6 | 523 | 9056333 | ||
| BC147301.1 | - | 6 | 523 | 9056336 | ||