Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290649.1 | NP_001277578.1 | 15, 16 | 1770 | ||
NM_029793.2 | NP_084069.1 | 16, 17 | 1845 | |||
GenBank mRNA | AK028516.1 | - | 16, 17 | 1765 | ||
AK044987.1 | - | 15, 16 | 2255 | |||
BC037037.1 | - | 11, 12 | 1181 | |||
BC043090.1 | - | 16, 17 | 1710 | 6418353 | ||
BC053068.1 | - | 15, 16 | 1698 | 6405595 | ||
BC080733.1 | - | 15, 16 | 1770 | 30110235 |