Search
Search

Abl1| Gene Symbol: Abl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112703.2 | NP_001106174.1 | 7 | 2458 | ||
| NM_001283045.1 | NP_001269974.1 | 7 | 1335 | |||
| NM_001283046.1 | NP_001269975.1 | 8 | 2663 | |||
| NM_001283047.1 | NP_001269976.1 | 8 | 2602 | |||
| NM_009594.4 | NP_033724.2 | 7 | 1229 | |||
| GenBank mRNA | AK088252.1 | - | 8 | 2588 | ||
| AK090095.1 | - | 7 | 1187 | |||
| AK166028.1 | - | 7 | 2447 | |||
| AK166210.1 | - | 8 | 2697 | |||
| AK190971.1 | - | 1 | 33 | |||
| AK218428.1 | - | 4 | 346 | |||
| BC059260.1 | - | 7 | 1256 | 6408990 | ||
| BC103770.1 | - | 7 | 1682 | 30848553 | ||
| CF727094.1 | - | 4 | 362 | |||
| CK626997.1 | - | 4 | 381 | |||
| J02995.1 | - | 7 | 1182 | |||
| L10656.1 | - | 5 | 783 | |||