Search
Search

Stmn2| Gene Symbol: Stmn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025285.2 | NP_079561.1 | 3 | 329 | ||
| GenBank mRNA | AK003659.1 | - | 3 | 329 | ||
| AK013020.1 | - | 3 | 330 | |||
| AK043202.1 | - | 3 | 342 | |||
| AK149212.1 | - | 3 | 354 | |||
| BC026538.1 | - | 3 | 291 | 4981703 | ||