Search
Search

Cetn2| Gene Symbol: Cetn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019405.6 | NP_062278.2 | 5 | 515 | ||
| GenBank mRNA | AK004081.1 | - | 5 | 516 | ||
| AK010541.1 | - | 5 | 506 | |||
| AK088488.1 | - | 3 | 347 | |||
| AK140156.1 | - | 5 | 533 | |||
| AK151033.1 | - | 5 | 583 | |||
| BC013545.1 | - | 4 | 499 | 4242430 | ||