Search
Search

Insig1| Gene Symbol: Insig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153526.5 | NP_705746.1 | 1 | 238 | ||
| GenBank mRNA | AF527630.1 | - | 1 | 252 | ||
| AK088380.1 | - | 1 | 285 | |||
| AK135941.1 | - | 1 | 289 | |||
| AK135989.1 | - | 1 | 290 | |||
| AK139913.1 | - | 1 | 288 | |||
| BC079559.1 | - | 1 | 238 | |||
| BC132167.1 | - | 1 | 246 | 40130444 | ||
| BC132499.1 | - | 1 | 246 | 40130776 | ||