Search
Search

Fars2| Gene Symbol: Fars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039189.2 | NP_001034278.1 | 3 | 633 | ||
| NM_024274.3 | NP_077236.1 | 4 | 1266 | |||
| GenBank mRNA | AK013211.1 | - | 4 | 1005 | ||
| AK020164.1 | - | 3 | 419 | |||
| AK054370.1 | - | 1032 | ||||
| AK076259.1 | - | 4 | 1050 | |||
| AK170986.1 | - | 4 | 1049 | |||
| BC002147.1 | - | 4 | 1036 | 3484842 | ||