Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cactin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027381.2 | NP_081657.1 | 3 | 1025 | ||
GenBank mRNA | AY934504.1 | - | 3 | 931 | ||
BC150986.1 | - | 3 | 1025 | 9087899 | ||
BC151180.1 | - | 3 | 1025 | 9088069 |