Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028182.1 | NP_082458.1 | 3 | 789 | ||
GenBank mRNA | AK008803.1 | - | 3 | 575 | ||
AK157396.1 | - | 4 | 587 | |||
AX776203.1 | - | 2 | 246 | |||
BC023784.1 | - | 3 | 182 | |||
BC116682.1 | - | 3 | 497 | 40106612 |