Search
Search

Mesdc1| Gene Symbol: Mesdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030705.4 | NP_109630.1 | 1 | 1802 | ||
| GenBank mRNA | AK036338.1 | - | 1768 | |||
| AK084510.1 | - | 1 | 1748 | |||
| AK089082.1 | - | 1 | 1757 | |||
| AK162446.1 | - | 1 | 1689 | |||
| BC018326.1 | - | 1 | 811 | 4018100 | ||