Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130421.2 | NP_569105.1 | 20, 21 | 1626 | ||
GenBank mRNA | AB072980.1 | - | 20, 21 | 1466 | ||
BC100066.1 | - | 20, 21 | 1626 | 7387128 |