Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIK3CB SIRNA Assays ›
          Gene Symbol:
          PIK3CB
          Gene Name:
          phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta
          Gene Aliases:
          P110BETA, PI3K, PI3KBETA, PIK3C1
          Chromosome Location:
          Chr.3: 138652698 - 138834938 on Build GRCh38
          UniGene ID:
          Hs.239818
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID VHS41143
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIK3CB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256045.1 NP_001242974.1 8 1352
          NM_006219.2 NP_006210.1 17 2486
          XM_005247530.2 XP_005247587.1 19 2835
          XM_006713659.3 XP_006713722.1 18 2706
          XM_011512895.2 XP_011511197.1 20 2831
          XM_011512896.1 XP_011511198.1 15 2100
          XM_017006619.1 XP_016862108.1 18 2509
          XM_017006620.1 XP_016862109.1 17 2256
          XM_017006621.1 XP_016862110.1 16 2205
          GenBank mRNA AK293754.1 - 8 1352
          AK302937.1 - 10 1365
          BC003393.1 - 2 119
          BC114432.1 - 17 2474 40008544
          CR749357.1 - 10 1071
          S67334.1 - 17 2470

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          endothelial cell proliferation
          regulation of cell-matrix adhesion
          phosphatidylinositol biosynthetic process
          cellular calcium ion homeostasis
          autophagy
          chemotaxis
          homophilic cell adhesion via plasma membrane adhesion molecules
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of autophagy
          positive regulation of gene expression
          phosphatidylinositol 3-kinase signaling
          regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          platelet activation
          phosphatidylinositol-3-phosphate biosynthetic process
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          embryonic cleavage
          phosphatidylinositol phosphorylation
          vascular endothelial growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          T cell receptor signaling pathway
          leukocyte migration
          angiogenesis involved in wound healing
          platelet aggregation
          regulation of clathrin-mediated endocytosis
          protein binding
          ATP binding
          1-phosphatidylinositol-3-kinase activity
          phosphatidylinositol 3-kinase activity
          1-phosphatidylinositol-4-phosphate 3-kinase activity
          insulin receptor substrate binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0