Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          SNHG3
          Gene Name:
          small nucleolar RNA host gene 3
          Gene Aliases:
          NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
          Chromosome Location:
          Chr.1: 28505943 - 28510892 on Build GRCh38
          UniGene ID:
          Hs.469723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258487

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002909.2
          3 456
          NR_036473.1
          4 564
          GenBank mRNA AJ006834.1 - LIT1316
          U17HG
          3 356
          AJ006835.1 - LIT1315
          U17HG
          3 356
          AK092726.1 -
          2 2089
          AK311577.1 -
          1 109
          BC006790.1 -
          3 338
          BC012122.1 -
          3 344
          BC017334.2 -
          3 331
          DA044266.1 -
          3 415
          DA135739.1 -
          4 527
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA820174.1 - 3 353 Chr. 1 - 28506046 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY075800.1 - 3 356 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA AA352342.1 - 2 307 Chr. 1 - 28508136 - 28509153 [+] on Build GRCh38
          GenBankmRNA HY180673.1 - 3 354 Chr. 1 - 28506045 - 28509192 [+] on Build GRCh38
          GenBankmRNA HY147758.1 - 3 361 Chr. 1 - 28506045 - 28509206 [+] on Build GRCh38
          GenBankmRNA BM450342.1 - 333 Chr. - - [-] on Build
          GenBankmRNA DA998284.1 - 3 364 Chr. 1 - 28506045 - 28509318 [+] on Build GRCh38
          GenBankmRNA CN366542.1 - 4 293 Chr. 1 - 28506055 - 28509581 [+] on Build GRCh38
          GenBankmRNA DA955189.1 - 3 358 Chr. 1 - 28506043 - 28509267 [+] on Build GRCh38
          GenBankmRNA DA933716.1 - 1 290 Chr. 1 - 28508824 - 28509359 [+] on Build GRCh38
          GenBankmRNA DB113121.1 - 3 356 Chr. 1 - 28506044 - 28509329 [+] on Build GRCh38
          GenBankmRNA BF941702.1 - 3 348 Chr. 1 - 28506050 - 28509152 [+] on Build GRCh38
          GenBankmRNA CN366538.1 - 3 356 Chr. 1 - 28506055 - 28509488 [+] on Build GRCh38
          GenBankmRNA HY182836.1 - 3 354 Chr. 1 - 28506045 - 28509242 [+] on Build GRCh38
          GenBankmRNA DA928567.1 - 3 353 Chr. 1 - 28506047 - 28509337 [+] on Build GRCh38
          GenBankmRNA BF795420.1 - 342 Chr. - - [-] on Build
          GenBankmRNA HY112100.1 - 3 430 Chr. 1 - 28505969 - 28509173 [+] on Build GRCh38
          GenBankmRNA CB985655.1 - 3 359 Chr. 1 - 28506043 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY059835.1 - 3 352 Chr. 1 - 28506045 - 28509325 [+] on Build GRCh38
          GenBankmRNA DA969742.1 - 3 360 Chr. 1 - 28506045 - 28509315 [+] on Build GRCh38
          GenBankmRNA DA968918.1 - 3 355 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BM802102.1 - 343 Chr. - - [-] on Build
          GenBankmRNA BM790281.1 - 3 343 Chr. 1 - 28506056 - 28509323 [+] on Build GRCh38
          GenBankmRNA AV763089.1 - 2 349 Chr. 1 - 28508127 - 28509528 [+] on Build GRCh38
          GenBankmRNA CD048541.1 - 3 375 Chr. 1 - 28506045 - 28509614 [+] on Build GRCh38
          GenBankmRNA DA478694.1 - 3 355 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA DA089660.1 - 3 416 Chr. 1 - 28505984 - 28509274 [+] on Build GRCh38
          GenBankmRNA HY176129.1 - 3 354 Chr. 1 - 28506045 - 28509283 [+] on Build GRCh38
          GenBankmRNA EB387088.1 - 1 17 Chr. 1 - 28509094 - 28509634 [+] on Build GRCh38
          GenBankmRNA DA704998.1 - 3 355 Chr. 1 - 28506044 - 28509252 [+] on Build GRCh38
          GenBankmRNA CN366541.1 - 2 312 Chr. 1 - 28508130 - 28509185 [+] on Build GRCh38
          GenBankmRNA DA728986.1 - 3 355 Chr. 1 - 28506044 - 28509467 [+] on Build GRCh38
          GenBankmRNA BQ224438.1 - 3 355 Chr. 1 - 28506055 - 28509617 [+] on Build GRCh38
          GenBankmRNA BE867363.1 - 2 329 Chr. 1 - 28508131 - 28509511 [+] on Build GRCh38
          GenBankmRNA DB024065.1 - 1 109 Chr. 1 - 28509002 - 28509575 [+] on Build GRCh38
          GenBankmRNA DA918003.1 - 3 356 Chr. 1 - 28506044 - 28509337 [+] on Build GRCh38
          GenBankmRNA DA817644.1 - 3 356 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA847021.1 - 3 356 Chr. 1 - 28506044 - 28509325 [+] on Build GRCh38
          GenBankmRNA HY177570.1 - 3 354 Chr. 1 - 28506045 - 28509225 [+] on Build GRCh38
          GenBankmRNA BM771983.1 - 3 355 Chr. 1 - 28506044 - 28509374 [+] on Build GRCh38
          GenBankmRNA BU187480.1 - 2 328 Chr. 1 - 28508127 - 28509634 [+] on Build GRCh38
          GenBankmRNA BI772556.1 - 2 338 Chr. 1 - 28508116 - 28509484 [+] on Build GRCh38
          GenBankmRNA DA961704.1 - 3 356 Chr. 1 - 28506044 - 28509303 [+] on Build GRCh38
          GenBankmRNA HY179875.1 - 3 353 Chr. 1 - 28506045 - 28509263 [+] on Build GRCh38
          GenBankmRNA DA922155.1 - 3 356 Chr. 1 - 28506044 - 28509320 [+] on Build GRCh38
          GenBankmRNA AV764560.2 - 3 349 Chr. 1 - 28506051 - 28509412 [+] on Build GRCh38
          GenBankmRNA DB157161.1 - 3 357 Chr. 1 - 28506043 - 28509332 [+] on Build GRCh38
          GenBankmRNA DA644956.1 - 3 356 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA BM479709.1 - 3 341 Chr. 1 - 28506059 - 28509634 [+] on Build GRCh38
          GenBankmRNA CN366536.1 - 2 310 Chr. 1 - 28508133 - 28509423 [+] on Build GRCh38
          GenBankmRNA HY173512.1 - 3 355 Chr. 1 - 28506045 - 28509232 [+] on Build GRCh38
          GenBankmRNA DA925005.1 - 3 355 Chr. 1 - 28506044 - 28509319 [+] on Build GRCh38
          GenBankmRNA BI833284.1 - 3 335 Chr. 1 - 28506060 - 28509531 [+] on Build GRCh38
          GenBankmRNA BU183513.1 - 3 343 Chr. 1 - 28506056 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM788818.1 - 2 332 Chr. 1 - 28508127 - 28509339 [+] on Build GRCh38
          GenBankmRNA DA936462.1 - 3 355 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DB112428.1 - 3 356 Chr. 1 - 28506044 - 28509353 [+] on Build GRCh38
          GenBankmRNA HY113224.1 - 3 412 Chr. 1 - 28505989 - 28509206 [+] on Build GRCh38
          GenBankmRNA CN366544.1 - 2 325 Chr. 1 - 28508127 - 28509283 [+] on Build GRCh38
          GenBankmRNA AL523116.3 - 2 327 Chr. 1 - 28508127 - 28509940 [+] on Build GRCh38
          GenBankmRNA DA952260.1 - 3 356 Chr. 1 - 28506044 - 28509313 [+] on Build GRCh38
          GenBankmRNA BE841288.1 - 1 42 Chr. 1 - 28509097 - 28509254 [+] on Build GRCh38
          GenBankmRNA BU943197.1 - 3 359 Chr. 1 - 28506044 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM834932.1 - 1 234 Chr. 1 - 28508876 - 28509340 [+] on Build GRCh38
          GenBankmRNA HY153255.1 - 3 354 Chr. 1 - 28506045 - 28509198 [+] on Build GRCh38
          GenBankmRNA CN366540.1 - 2 340 Chr. 1 - 28508127 - 28509421 [+] on Build GRCh38
          GenBankmRNA DA565956.1 - 3 355 Chr. 1 - 28506044 - 28509324 [+] on Build GRCh38
          GenBankmRNA HY084156.1 - 3 354 Chr. 1 - 28506045 - 28509217 [+] on Build GRCh38
          GenBankmRNA DB128729.1 - 3 355 Chr. 1 - 28506044 - 28509326 [+] on Build GRCh38
          GenBankmRNA BG474710.1 - 3 346 Chr. 1 - 28506055 - 28509618 [+] on Build GRCh38
          GenBankmRNA CB157773.1 - 3 344 Chr. 1 - 28506055 - 28509221 [+] on Build GRCh38
          GenBankmRNA DB262011.1 - 3 356 Chr. 1 - 28506044 - 28509330 [+] on Build GRCh38
          GenBankmRNA DB271395.1 - 3 356 Chr. 1 - 28506044 - 28509295 [+] on Build GRCh38
          GenBankmRNA DA943290.1 - 3 357 Chr. 1 - 28506043 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY089919.1 - 3 354 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA CB963044.1 - 3 410 Chr. 1 - 28505990 - 28509462 [+] on Build GRCh38
          GenBankmRNA HY179907.1 - 3 354 Chr. 1 - 28506045 - 28509261 [+] on Build GRCh38
          GenBankmRNA DA510712.1 - 3 356 Chr. 1 - 28506044 - 28509216 [+] on Build GRCh38
          GenBankmRNA BM759606.1 - 3 356 Chr. 1 - 28506043 - 28509424 [+] on Build GRCh38
          GenBankmRNA BF982554.1 - 2 303 Chr. 1 - 28508137 - 28509570 [+] on Build GRCh38
          GenBankmRNA CN366535.1 - 3 346 Chr. 1 - 28506053 - 28509489 [+] on Build GRCh38
          GenBankmRNA DA787816.1 - 3 355 Chr. 1 - 28506044 - 28509270 [+] on Build GRCh38
          GenBankmRNA BG023899.1 - 326 Chr. - - [-] on Build
          GenBankmRNA DA902617.1 - 3 355 Chr. 1 - 28506045 - 28509297 [+] on Build GRCh38
          GenBankmRNA HY176989.1 - 3 353 Chr. 1 - 28506045 - 28509229 [+] on Build GRCh38
          GenBankmRNA HY175139.1 - 3 354 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA CN366543.1 - 3 353 Chr. 1 - 28506055 - 28509366 [+] on Build GRCh38
          GenBankmRNA BY796950.2 - 2 324 Chr. 1 - 28508127 - 28509414 [+] on Build GRCh38
          GenBankmRNA DA381375.1 - 1 399 Chr. 1 - 28508764 - 28509340 [+] on Build GRCh38
          GenBankmRNA HY182945.1 - 3 354 Chr. 1 - 28506045 - 28509230 [+] on Build GRCh38
          GenBankmRNA CN366539.1 - 3 331 Chr. 1 - 28506068 - 28509242 [+] on Build GRCh38
          GenBankmRNA DA932306.1 - 3 355 Chr. 1 - 28506044 - 28509327 [+] on Build GRCh38
          GenBankmRNA BG028447.1 - 3 341 Chr. 1 - 28506057 - 28509522 [+] on Build GRCh38
          GenBankmRNA CB128113.1 - 3 358 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA BG291633.1 - 3 337 Chr. 1 - 28506067 - 28509489 [+] on Build GRCh38
          GenBankmRNA CB163464.1 - 3 339 Chr. 1 - 28506060 - 28509328 [+] on Build GRCh38
          GenBankmRNA AI004003.1 - 1 188 Chr. 1 - 28508923 - 28509326 [+] on Build GRCh38
          GenBankmRNA BI833122.1 - 3 338 Chr. 1 - 28506059 - 28509542 [+] on Build GRCh38
          GenBankmRNA DA224254.1 - 3 357 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA926278.1 - 3 356 Chr. 1 - 28506044 - 28509334 [+] on Build GRCh38
          GenBankmRNA DA056610.1 - 3 415 Chr. 1 - 28505984 - 28509252 [+] on Build GRCh38
          GenBankmRNA BU198833.1 - 3 359 Chr. 1 - 28506044 - 28509422 [+] on Build GRCh38
          GenBankmRNA BM802666.1 - 343 Chr. - - [-] on Build
          GenBankmRNA BF036396.1 - 2 321 Chr. 1 - 28508127 - 28509376 [+] on Build GRCh38
          GenBankmRNA DA652687.1 - 3 357 Chr. 1 - 28506044 - 28509333 [+] on Build GRCh38
          GenBankmRNA BQ218931.1 - 3 339 Chr. 1 - 28506060 - 28509544 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-v2dhc:80/100.66.79.56:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0