Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CA5BP1 SIRNA Assays ›
          Gene Symbol:
          CA5BP1 CA5B
          Gene Name:
          carbonic anhydrase 5B pseudogene 1
          carbonic anhydrase 5B
          Gene Aliases:
          CA5BL, CA5BP, PRO2325
          CA-VB
          Chromosome Location:
          Chr.X: 15674916 - 15703351 on Build GRCh38
          Chromosome Location:
          Chr.X: 15738281 - 15787625 on Build GRCh38
          UniGene ID:
          Hs.732244
          Hs.653287
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n265035

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CA5BP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026551.1 3, 4 327
          GenBank mRNA CD675860.1 - 3, 4 336
          DB271346.1 - 2, 3 242
          Gene Symbol: CA5B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017029249.1 XP_016884738.1 1, 2 102
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX345337.2 - 3, 4 343 Chr. X - 15676294 - 15697460 [+] on Build GRCh38
          GenBankmRNA CN408436.1 - 2, 3 496 Chr. X - 15675480 - 15697264 [+] on Build GRCh38
          GenBankmRNA AA279513.1 - 2, 3 263 Chr. X - 15675709 - 15702883 [+] on Build GRCh38
          GenBankmRNA AA459755.1 - 2, 3 217 Chr. X - 15647141 - 15702860 [+] on Build GRCh38
          GenBankmRNA BI765364.1 - 2, 3 225 Chr. X - 15675546 - 15703349 [+] on Build GRCh38
          GenBankmRNA DR000956.1 - 2, 3 261 Chr. X - 15675725 - 15697515 [+] on Build GRCh38
          GenBankmRNA BI333715.1 - 2, 3 234 Chr. X - 15674932 - 15697799 [+] on Build GRCh38
          GenBankmRNA BQ919066.1 - 2, 3 258 Chr. X - 15675718 - 15703339 [+] on Build GRCh38
          GenBankmRNA BM906941.1 - 273 Chr. - - [-] on Build
          GenBankmRNA BI334916.1 - 2, 3 234 Chr. X - 15674932 - 15697884 [+] on Build GRCh38
          GenBankmRNA BU541060.1 - 633 Chr. - - [-] on Build
          GenBankmRNA BQ439983.1 - 2, 3 263 Chr. X - 15675713 - 15697743 [+] on Build GRCh38
          GenBankmRNA BM045733.1 - 2, 3 480 Chr. X - 15675496 - 15697432 [+] on Build GRCh38
          GenBankmRNA BM044609.1 - 2, 3 480 Chr. X - 15675496 - 15697532 [+] on Build GRCh38
          GenBankmRNA BI833900.1 - 2, 3 238 Chr. X - 15674984 - 15703228 [+] on Build GRCh38
          GenBankmRNA BU542020.1 - 2, 3 478 Chr. X - 15675498 - 15697428 [+] on Build GRCh38
          GenBankmRNA BM550787.1 - 2, 3 277 Chr. X - 15675702 - 15697819 [+] on Build GRCh38
          GenBankmRNA BM046212.1 - 2, 3 480 Chr. X - 15675496 - 15697522 [+] on Build GRCh38
          GenBankmRNA DB288220.1 - 2, 3 230 Chr. X - 15674924 - 15697463 [+] on Build GRCh38
          GenBankmRNA CF242929.1 - 1, 2 125 Chr. X - 15688734 - 15702531 [+] on Build GRCh38
          GenBankmRNA BI917660.1 - 2, 3 257 Chr. X - 15675719 - 15703185 [+] on Build GRCh38
          GenBankmRNA DN998494.1 - 2, 3 259 Chr. X - 15675719 - 15697648 [+] on Build GRCh38
          GenBankmRNA BG828555.1 - 2, 3 243 Chr. X - 15675735 - 15703267 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          one-carbon metabolic process
          bicarbonate transport
          carbonate dehydratase activity
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0