Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHRM3 SIRNA Assays ›
          Gene Symbol:
          CHRM3
          Gene Name:
          cholinergic receptor muscarinic 3
          Gene Aliases:
          EGBRS, HM3, PBS
          Chromosome Location:
          Chr.1: 239386565 - 239915450 on Build GRCh38
          UniGene ID:
          Hs.155736
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n320638
          Size
          Availability Made To Order
          Catalog #
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHRM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005273032.3 XP_005273089.1 6 8543
          XM_011544041.2 XP_011542343.1 7 8642
          XM_011544043.2 XP_011542345.1 9 8975
          XM_011544044.2 XP_011542346.1 8 8746
          XM_011544045.2 XP_011542347.1 8 8871
          XM_011544046.2 XP_011542348.1 6 8579
          XM_011544047.2 XP_011542349.1 6 8515
          XM_017000152.1 XP_016855641.1 8 11568
          XM_017000153.1 XP_016855642.1 7 8646
          XM_017000154.1 XP_016855643.1 6 8340
          XM_017000155.1 XP_016855644.1 7 8683
          XM_017000156.1 XP_016855645.1 7 8647
          XM_017000157.1 XP_016855646.1 5 8248
          XM_017000158.1 XP_016855647.1 5 8480
          XM_017000159.1 XP_016855648.1 5 8452
          XM_017000160.1 XP_016855649.1 4 9302
          XM_017000161.1 XP_016855650.1 6 8584
          XM_017000162.1 XP_016855651.1 5 8416
          XM_017000163.1 XP_016855652.1 4 12278
          GenBank mRNA AK095715.1 - 1 3248
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CD678044.1 - 1 160 Chr. 1 - 239914751 - 239915381 [+] on Build GRCh38
          GenBankmRNA EL952133.1 - 1 140 Chr. 1 - 239914821 - 239915303 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of vascular smooth muscle contraction
          cellular protein modification process
          smooth muscle contraction
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled acetylcholine receptor signaling pathway
          phospholipase C-activating G-protein coupled acetylcholine receptor signaling pathway
          G-protein coupled acetylcholine receptor signaling pathway
          synaptic transmission, cholinergic
          nervous system development
          cell proliferation
          positive regulation of smooth muscle contraction
          saliva secretion
          phosphatidylinositol phospholipase C activity
          receptor activity
          protein binding
          drug binding
          G-protein coupled acetylcholine receptor activity
          acetylcholine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-kkgv7:80/100.66.73.110:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0