Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INHBA SIRNA Assays ›
          Gene Symbol:
          INHBA
          Gene Name:
          inhibin beta A subunit
          Gene Aliases:
          EDF, FRP
          Chromosome Location:
          Chr.7: 41685101 - 41710532 on Build GRCh38
          UniGene ID:
          Hs.28792
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n341385

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INHBA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017012174.1 XP_016867663.1 5 6721
          XM_017012175.1 XP_016867664.1 3 6405
          XM_017012176.1 XP_016867665.1 5 6589
          GenBank mRNA AK001903.1 - 1 697

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          ovarian follicle development
          hair follicle development
          hematopoietic progenitor cell differentiation
          regulation of transcription from RNA polymerase II promoter
          defense response
          cell cycle arrest
          cell surface receptor signaling pathway
          cell-cell signaling
          nervous system development
          negative regulation of cell proliferation
          male gonad development
          positive regulation of gene expression
          positive regulation of pathway-restricted SMAD protein phosphorylation
          striatal medium spiny neuron differentiation
          cell differentiation
          erythrocyte differentiation
          negative regulation of cell growth
          positive regulation of cellular protein metabolic process
          activin receptor signaling pathway
          endodermal cell differentiation
          growth
          negative regulation of phosphorylation
          odontogenesis
          response to drug
          hemoglobin biosynthetic process
          progesterone secretion
          regulation of MAPK cascade
          negative regulation of interferon-gamma biosynthetic process
          negative regulation of B cell differentiation
          positive regulation of erythrocyte differentiation
          negative regulation of macrophage differentiation
          negative regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of follicle-stimulating hormone secretion
          positive regulation of follicle-stimulating hormone secretion
          negative regulation of follicle-stimulating hormone secretion
          mesodermal cell differentiation
          cell development
          palate development
          positive regulation of ovulation
          SMAD protein signal transduction
          eyelid development in camera-type eye
          cellular response to follicle-stimulating hormone stimulus
          cellular response to cholesterol
          GABAergic neuron differentiation
          extrinsic apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          cytokine activity
          transforming growth factor beta receptor binding
          hormone activity
          protein binding
          growth factor activity
          peptide hormone binding
          inhibin binding
          identical protein binding
          protein heterodimerization activity
          type II activin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0