Search
Search

DLGAP1| Gene Symbol: DLGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003809.2 | NP_001003809.1 | 10 | 4573 | ||
| NM_001242762.1 | NP_001229691.1 | 10 | 4516 | |||
| NM_001242763.1 | NP_001229692.1 | 11 | 4630 | |||
| NM_001242764.1 | NP_001229693.1 | 10 | 4546 | |||
| NM_001242766.1 | NP_001229695.1 | 11 | 4603 | |||
| NM_001308390.1 | NP_001295319.1 | 11 | 4628 | |||
| NM_004746.3 | NP_004737.2 | 13 | 5930 | |||
| XM_005258171.2 | XP_005258228.1 | 14 | 6001 | |||
| XM_005258172.1 | XP_005258229.1 | 14 | 5996 | |||
| XM_005258173.3 | XP_005258230.2 | 12 | 4792 | |||
| XM_005258174.4 | XP_005258231.1 | 11 | 4849 | |||
| XM_006722367.3 | XP_006722430.1 | 12 | 4960 | |||
| XM_017026080.1 | XP_016881569.1 | 11 | 4929 | |||
| XM_017026081.1 | XP_016881570.1 | 10 | 4593 | |||
| XM_017026083.1 | XP_016881572.1 | 10 | 4818 | |||
| GenBank mRNA | AK095761.1 | - | 1 | 2136 | ||