Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ELDR SIRNA Assays ›
          Gene Symbol:
          Eldr
          Gene Name:
          Egfr long non-coding downstream RNA
          Gene Aliases:
          2810442I21Rik, 4930500E12Rik, Fabl
          Chromosome Location:
          Chr.11: 16934709 - 16951282 on Build GRCm38
          UniGene ID:
          Mm.268598
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n519609

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eldr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_110421.1 1, 2 265
          NR_110422.1 1, 2 265
          NR_110424.1 1, 2 265
          NR_110425.1 1, 2 265
          GenBank mRNA AK013290.1 - 1, 2 107
          AK015657.1 - 1, 2 266
          AK165613.1 - 1, 2 96
          BC037766.1 - 1, 2 82
          BC054406.1 - 1, 2 26
          BI990615.1 - 1, 2 65
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY733051.1 - 1, 2 95 Chr. 11 - 16935614 - 16951109 [-] on Build GRCm38
          GenBankmRNA BY042125.1 - 1, 2 73 Chr. 11 - 16935701 - 16951088 [-] on Build GRCm38
          GenBankmRNA BY041527.1 - 1, 2 73 Chr. 11 - 16935630 - 16951088 [-] on Build GRCm38
          GenBankmRNA W53843.1 - 92 Chr. - - [-] on Build
          GenBankmRNA BY039667.1 - 1, 2 73 Chr. 11 - 16935705 - 16951088 [-] on Build GRCm38
          GenBankmRNA BE332295.1 - 1, 2 82 Chr. 11 - 16936301 - 16951099 [-] on Build GRCm38
          GenBankmRNA BY050622.1 - 1, 2 73 Chr. 11 - 16935690 - 16951088 [-] on Build GRCm38
          GenBankmRNA BX523360.1 - 1, 2 83 Chr. 11 - 16936133 - 16951099 [-] on Build GRCm38
          GenBankmRNA BY755193.1 - 1, 2 97 Chr. 11 - 16935621 - 16951109 [-] on Build GRCm38
          GenBankmRNA AA739401.1 - 16 Chr. 11 - 16936178 - 16950935 [-] on Build GRCm38
          GenBankmRNA BU517449.1 - 1, 2 25 Chr. 11 - 16935315 - 16951042 [-] on Build GRCm38
          GenBankmRNA BB617529.1 - 1, 2 105 Chr. 11 - 16936063 - 16951122 [-] on Build GRCm38
          GenBankmRNA BY049998.1 - 1, 2 73 Chr. 11 - 16935678 - 16951088 [-] on Build GRCm38
          GenBankmRNA AA155535.1 - 1, 2 91 Chr. 11 - 16936426 - 16951108 [-] on Build GRCm38
          GenBankmRNA BY045022.1 - 1, 2 73 Chr. 11 - 16935689 - 16951088 [-] on Build GRCm38
          GenBankmRNA BY041151.1 - 1, 2 73 Chr. 11 - 16935692 - 16951089 [-] on Build GRCm38
          GenBankmRNA BM875981.1 - 1, 2 104 Chr. 11 - 16935716 - 16951098 [-] on Build GRCm38
          GenBankmRNA W97663.1 - 92 Chr. - - [-] on Build

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0