Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADAR SIRNA Assays ›
          Gene Symbol:
          ADAR
          Gene Name:
          adenosine deaminase, RNA specific
          Gene Aliases:
          ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
          Chromosome Location:
          Chr.1: 154582057 - 154628016 on Build GRCh38
          UniGene ID:
          Hs.12341
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1007
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADAR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025107.2 NP_001020278.1 2 1229
          NM_001111.4 NP_001102.2 2 1360
          NM_001193495.1 NP_001180424.1 2 1398
          NM_015840.3 NP_056655.2 2 1360
          NM_015841.3 NP_056656.2 2 1360
          XM_006711109.2 XP_006711172.1 2 1166
          XM_006711111.3 XP_006711174.1 3 1288
          XM_006711112.2 XP_006711175.1 3 990
          XM_006711113.2 XP_006711176.1 3 1064
          XM_011509060.1 XP_011507362.1 2 1262
          XM_011509061.2 XP_011507363.2 2 1229
          XM_011509062.1 XP_011507364.1 2 1189
          XM_017000037.1 XP_016855526.1 3 1157
          GenBank mRNA AB209891.1 - 2 1173
          AK097241.1 - 3 1000
          AK304153.1 - 2 1314
          BC038227.1 - 2 1287 5497548
          BX538232.1 - 2 1213
          BX640741.1 - 2 1211
          KJ544856.1 - 1 1118
          U10439.1 - 2 1272
          U18121.1 - 2 1165
          X79448.1 - 2 1398
          X79449.1 - 2 1320
          X98559.1 - 1 1103

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00241666_m1

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          in utero embryonic development
          hematopoietic progenitor cell differentiation
          somatic diversification of immune receptors via somatic mutation
          adenosine to inosine editing
          mRNA processing
          response to virus
          base conversion or substitution editing
          erythrocyte differentiation
          pre-miRNA processing
          miRNA loading onto RISC involved in gene silencing by miRNA
          response to interferon-alpha
          negative regulation of apoptotic process
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          positive regulation of viral genome replication
          negative regulation of viral genome replication
          innate immune response
          defense response to virus
          definitive hemopoiesis
          type I interferon signaling pathway
          negative regulation of type I interferon-mediated signaling pathway
          hematopoietic stem cell homeostasis
          cellular response to virus
          negative regulation of RNA interference
          DNA binding
          double-stranded RNA adenosine deaminase activity
          protein binding
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0