Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172643.5 | NP_766231.2 | 3 | 1324 | ||
GenBank mRNA | AK034756.1 | - | 2 | 423 | ||
AK036544.1 | - | 3 | 1323 | |||
AK163302.1 | - | 2 | 597 | |||
BC046465.1 | - | 2 | 493 | |||
BC150859.1 | - | 2 | 663 | 9087770 |