Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPM1A SIRNA Assays ›
          Gene Symbol:
          PPM1A
          Gene Name:
          protein phosphatase, Mg2+/Mn2+ dependent 1A
          Gene Aliases:
          PP2C-ALPHA, PP2CA, PP2Calpha
          Chromosome Location:
          Chr.14: 60245752 - 60299087 on Build GRCh38
          UniGene ID:
          Hs.130036
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10919
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 562.00
          Your Price
          Online offer: 533.65
          533.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPM1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021003.4 NP_066283.1 2 1228
          NM_177951.2 NP_808820.1 3 1330
          NM_177952.2 NP_808821.2 2 1093
          XM_005267779.1 XP_005267836.1 3 1175
          XM_005267781.1 XP_005267838.1 3 1333
          XM_011536872.1 XP_011535174.1 5 1595
          XM_011536876.1 XP_011535178.1 4 1452
          XM_011536877.1 XP_011535179.1 5 1495
          XM_011536878.1 XP_011535180.1 4 1418
          XM_011536879.2 XP_011535181.1 4 1287
          XM_011536880.2 XP_011535182.1 5 1341
          XM_011536881.2 XP_011535183.1 3 1201
          XM_011536882.2 XP_011535184.1 2 870
          XM_011536883.1 XP_011535185.1 3 1284
          XM_011536884.1 XP_011535186.1 2 841
          XM_017021381.1 XP_016876870.1 5 2465
          XM_017021382.1 XP_016876871.1 7 1715
          XM_017021383.1 XP_016876872.1 4 1351
          XM_017021384.1 XP_016876873.1 4 1043
          XM_017021385.1 XP_016876874.1 3 1318
          XM_017021386.1 XP_016876875.1 3 1081
          XM_017021387.1 XP_016876876.1 4 1327
          GenBank mRNA AB451247.1 - 1 778
          AB451371.1 - 1 778
          AF070670.1 - 3 1137
          AK097843.1 - 2 1093
          AK308225.1 - 2 955
          AK313337.1 - 3 1175
          BC026691.1 - 2 1081 3847638
          BC063243.1 - 2 1011 6011486
          S87759.1 - 2 1135

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017038, XM_006240195, XM_006240194, XM_006240193, J04503.
          This siRNA also targets the following transcripts in Mouse: NM_008910, BC008595, AF369981, AF259673, AK210783, and others.

          Matching Taqman Gene Expression Assay:


          Hs00221372_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein dephosphorylation
          N-terminal protein myristoylation
          cell cycle arrest
          negative regulation of SMAD protein complex assembly
          Wnt signaling pathway
          dephosphorylation
          positive regulation of Wnt signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          peptidyl-threonine dephosphorylation
          negative regulation of NF-kappaB import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription, DNA-templated
          positive regulation of protein export from nucleus
          cellular response to transforming growth factor beta stimulus
          magnesium ion binding
          protein serine/threonine phosphatase activity
          signal transducer activity
          protein binding
          manganese ion binding
          calmodulin-dependent protein phosphatase activity
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0