Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ROCK1 SIRNA Assays ›
          Gene Symbol:
          ROCK1
          Gene Name:
          Rho associated coiled-coil containing protein kinase 1
          Gene Aliases:
          P160ROCK, ROCK-I
          Chromosome Location:
          Chr.18: 20949740 - 21111851 on Build GRCh38
          UniGene ID:
          Hs.306307
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s12098
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ROCK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005406.2 NP_005397.1 18 2996
          XM_011526137.2 XP_011524439.1 7 1230
          GenBank mRNA AB208965.1 - 10 1089
          AK307388.1 - 18 2811
          BC113114.1 - 18 2071 7961989
          U43195.1 - 18 2055

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00178463_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          apical constriction
          protein phosphorylation
          cellular component disassembly involved in execution phase of apoptosis
          smooth muscle contraction
          leukocyte cell-cell adhesion
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          Rho protein signal transduction
          negative regulation of angiogenesis
          membrane to membrane docking
          actin cytoskeleton organization
          regulation of cell adhesion
          cortical actin cytoskeleton organization
          bleb assembly
          negative regulation of protein binding
          regulation of actin cytoskeleton organization
          negative regulation of myosin-light-chain-phosphatase activity
          negative regulation of neuron apoptotic process
          regulation of keratinocyte differentiation
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          leukocyte migration
          leukocyte tethering or rolling
          myoblast migration
          regulation of stress fiber assembly
          regulation of focal adhesion assembly
          positive regulation of focal adhesion assembly
          establishment of protein localization to plasma membrane
          regulation of establishment of endothelial barrier
          negative regulation of bicellular tight junction assembly
          regulation of establishment of cell polarity
          regulation of cell motility
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          GTP-Rho binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0