Search
Search

| Gene Symbol: RPL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000981.3 | NP_000972.1 | 4 | 385 | ||
| XM_005257564.3 | XP_005257621.1 | 4 | 734 | |||
| GenBank mRNA | AB019566.1 | - | 1 | 96 | ||
| AK130962.1 | - | 4 | 352 | |||
| AK223082.1 | - | 4 | 347 | |||
| AK311858.1 | - | 4 | 353 | |||
| AY157980.1 | - | 3 | 267 | |||
| BC000530.2 | - | 4 | 335 | 2961653 | ||
| BC013016.2 | - | 3 | 332 | 3010178 | ||
| BC062709.1 | - | 4 | 350 | 6457109 | ||
| BC066315.1 | - | 4 | 351 | 5287470 | ||
| BC095445.1 | - | 4 | 385 | 30721753 | ||
| BX537435.1 | - | 4 | 350 | |||
| S56985.1 | - | 1 | 334 | |||
| X63527.1 | - | 4 | 351 | |||
| Gene Symbol: RPL19P12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026660.1 | 1 | 787 | |||