Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ST3GAL3 SIRNA Assays ›
          Gene Symbol:
          ST3GAL3
          Gene Name:
          ST3 beta-galactoside alpha-2,3-sialyltransferase 3
          Gene Aliases:
          EIEE15, MRT12, SIAT6, ST3GALII, ST3GalIII, ST3N
          Chromosome Location:
          Chr.1: 43707497 - 43931165 on Build GRCh38
          UniGene ID:
          Hs.597915
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s12855

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ST3GAL3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270459.1 NP_001257388.1 2 277
          NM_001270460.1 NP_001257389.1 2 277
          NM_001270461.1 NP_001257390.1 2 277
          NM_001270462.1 NP_001257391.1 2 277
          NM_001270463.1 NP_001257392.1 2 277
          NM_001270464.1 NP_001257393.1 2 277
          NM_001270465.1 NP_001257394.1 2 277
          NM_001270466.1 NP_001257395.1 2 277
          NM_006279.3 NP_006270.1 2 277
          NM_174963.3 NP_777623.2 2 277
          NM_174964.2 NP_777624.1 2 277
          NM_174965.2 NP_777625.1 2 277
          NM_174966.2 NP_777626.1 2 277
          NM_174967.2 NP_777627.1 2 277
          NM_174968.3 NP_777628.2 2 277
          NM_174969.2 NP_777629.1 2 277
          NM_174970.2 NP_777630.1 2 277
          NM_174971.3 NP_777631.2 2 277
          NR_073016.1 2 277
          NR_073017.1 2 277
          NR_073018.1 2 277
          NR_073019.1 2 277
          NR_073020.1 2 277
          NR_073021.1 2 277
          NR_073023.1 2 277
          XM_006710825.3 XP_006710888.1 2 313
          XM_006710827.3 XP_006710890.1 2 279
          XM_006710828.3 XP_006710891.1 2 279
          XM_011541973.2 XP_011540275.1 2 279
          XM_011541974.2 XP_011540276.1 2 279
          XM_011541976.2 XP_011540278.1 2 279
          XM_011541977.2 XP_011540279.1 2 279
          XM_011541981.2 XP_011540283.1 2 279
          XM_011541983.2 XP_011540285.1 2 279
          XM_011541984.2 XP_011540286.1 2 279
          XM_011541985.2 XP_011540287.1 2 279
          XM_011541986.2 XP_011540288.1 2 279
          XM_011541987.2 XP_011540289.1 2 279
          XM_017002109.1 XP_016857598.1 2 342
          XM_017002110.1 XP_016857599.1 2 279
          XM_017002111.1 XP_016857600.1 1 116
          XM_017002112.1 XP_016857601.1 1 116
          XM_017002113.1 XP_016857602.1 2 357
          XM_017002114.1 XP_016857603.1 1 116
          XM_017002115.1 XP_016857604.1 2 279
          XM_017002116.1 XP_016857605.1 1 116
          XM_017002117.1 XP_016857606.1 2 279
          XM_017002118.1 XP_016857607.1 2 279
          XM_017002119.1 XP_016857608.1 2 277
          XM_017002120.1 XP_016857609.1 2 276
          XM_017002121.1 XP_016857610.1 1 117
          XM_017002122.1 XP_016857611.1 1 116
          GenBank mRNA AF425851.1 - 1 86
          AF425852.1 - 1 86
          AF425853.1 - 1 86
          AF425854.1 - 1 86
          AF425855.1 - 1 86
          AF425856.1 - 1 86
          AF425857.1 - 1 86
          AF425858.1 - 1 86
          AF425859.1 - 1 86
          AF425860.1 - 1 86
          AF425861.1 - 1 86
          AF425862.1 - 1 86
          AF425863.1 - 1 86
          AF425864.1 - 1 86
          AF425865.1 - 1 86
          AF425866.1 - 1 86
          AF425867.1 - 1 86
          AF425868.1 - 1 86
          AF425869.1 - 1 86
          AK301829.1 - 2 263
          AK310265.1 - 2 291
          AY051143.1 - 2 262
          AY167992.1 - 1 86
          AY167993.1 - 1 86
          AY167994.1 - 1 86
          AY167995.1 - 1 86
          AY167996.1 - 1 86
          AY167997.1 - 1 86
          AY167998.1 - 1 86
          BC050380.1 - 2 261 5769575
          L23768.1 - 1 86

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          ganglioside biosynthetic process
          oligosaccharide metabolic process
          O-glycan processing
          keratan sulfate biosynthetic process
          protein N-linked glycosylation via asparagine
          sialylation
          beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
          N-acetyllactosaminide alpha-2,3-sialyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0