Search
Search

SNRPF| Gene Symbol: SNRPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003095.2 | NP_003086.1 | 2 | 240 | ||
| GenBank mRNA | AK311752.1 | - | 2 | 203 | ||
| BC002505.2 | - | 2 | 233 | 3051263 | ||
| BC063397.1 | - | 2 | 190 | 5176590 | ||
| BC128452.1 | - | 2 | 143 | 40123237 | ||
| BC128453.1 | - | 2 | 142 | 40123239 | ||
| BU861031.1 | - | 2 | 243 | |||
| CR456751.1 | - | 1 | 97 | |||
| X85372.1 | - | 2 | 212 | |||