Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130430.1 | NP_569114.1 | 3 | 748 | ||
GenBank mRNA | AF067728.1 | - | 3 | 748 | ||
FQ214178.1 | - | 292 | ||||
FQ225583.1 | - | 281 | ||||
FQ229304.1 | - | 314 |
Gene Symbol: LOC103694906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773903.1 | XP_008772125.1 | 2 | 485 |