Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNF SIRNA Assays ›
Gene Symbol:
TNF
Gene Name:
tumor necrosis factor
Gene Aliases:
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
Chromosome Location:
Chr.6: 31575567 - 31578336 on Build GRCh38
UniGene ID:
Hs.241570
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14248
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000594.3
NP_000585.2
4
1217
GenBank mRNA
BC028148.1
-
4
1199
5217636
M10988.1
-
4
1127
X01394.1
-
4
1194
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
activation of MAPKKK activity
activation of MAPK activity
positive regulation of cytokine production
positive regulation of protein phosphorylation
chronic inflammatory response to antigenic stimulus
negative regulation of cytokine secretion involved in immune response
positive regulation of chronic inflammatory response to antigenic stimulus
positive regulation of humoral immune response mediated by circulating immunoglobulin
glucose metabolic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
inflammatory response
humoral immune response
I-kappaB kinase/NF-kappaB signaling
JNK cascade
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
response to virus
response to salt stress
positive regulation of gene expression
negative regulation of gene expression
negative regulation of alkaline phosphatase activity
regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of lipid storage
extracellular matrix organization
osteoclast differentiation
sequestering of triglyceride
cortical actin cytoskeleton organization
positive regulation of protein complex assembly
positive regulation of fever generation
lipopolysaccharide-mediated signaling pathway
negative regulation of interleukin-6 production
positive regulation of chemokine production
positive regulation of interferon-gamma production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
receptor biosynthetic process
positive regulation of peptidyl-serine phosphorylation
tumor necrosis factor-mediated signaling pathway
positive regulation of heterotypic cell-cell adhesion
negative regulation of myosin-light-chain-phosphatase activity
positive regulation of NF-kappaB import into nucleus
positive regulation of apoptotic process
positive regulation of programmed cell death
regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of protein complex disassembly
positive regulation of protein complex disassembly
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAP kinase activity
protein kinase B signaling
positive regulation of JUN kinase activity
negative regulation of growth of symbiont in host
negative regulation of viral genome replication
positive regulation of chemokine biosynthetic process
positive regulation of interleukin-8 biosynthetic process
positive regulation of nitric oxide biosynthetic process
negative regulation of fat cell differentiation
negative regulation of myoblast differentiation
negative regulation of osteoblast differentiation
positive regulation of osteoclast differentiation
positive regulation of cell adhesion
positive regulation of protein kinase activity
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of translational initiation by iron
negative regulation of glucose import
embryonic digestive tract development
positive regulation of smooth muscle cell proliferation
positive regulation of cytokine secretion
positive regulation of phagocytosis
regulation of insulin secretion
defense response to Gram-positive bacterium
leukocyte tethering or rolling
negative regulation of lipid catabolic process
regulation of immunoglobulin secretion
positive regulation of membrane protein ectodomain proteolysis
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
positive regulation of protein transport
response to glucocorticoid
positive regulation of NFAT protein import into nucleus
positive regulation of hair follicle development
positive regulation of protein kinase B signaling
positive regulation of vitamin D biosynthetic process
positive regulation of calcidiol 1-monooxygenase activity
epithelial cell proliferation involved in salivary gland morphogenesis
regulation of branching involved in salivary gland morphogenesis
negative regulation of branching involved in lung morphogenesis
positive regulation of ERK1 and ERK2 cascade
cellular response to amino acid stimulus
cellular response to nicotine
cellular response to organic cyclic compound
death-inducing signaling complex assembly
positive regulation of mononuclear cell migration
positive regulation of podosome assembly
establishment of protein localization to plasma membrane
extrinsic apoptotic signaling pathway
necroptotic signaling pathway
positive regulation of NIK/NF-kappaB signaling
positive regulation of superoxide dismutase activity
regulation of establishment of endothelial barrier
negative regulation of bicellular tight junction assembly
positive regulation of leukocyte adhesion to vascular endothelial cell
positive regulation of leukocyte adhesion to arterial endothelial cell
positive regulation of protein localization to cell surface
positive regulation of ceramide biosynthetic process
positive regulation of blood microparticle formation
positive regulation of chemokine (C-X-C motif) ligand 2 production
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protease binding
cytokine activity
tumor necrosis factor receptor binding
protein binding
identical protein binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0