Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBE2V1 SIRNA Assays ›
          Gene Symbol:
          UBE2V1 TMEM189-UBE2V1
          Gene Name:
          ubiquitin conjugating enzyme E2 V1
          TMEM189-UBE2V1 readthrough
          Gene Aliases:
          CIR1, CROC-1, CROC1, UBE2V, UEV-1, UEV1, UEV1A
          CROC-1B, CROC1B, KUA-UEV
          Chromosome Location:
          Chr.20: 50081124 - 50115959 on Build GRCh38
          Chromosome Location:
          Chr.20: 50081124 - 50153798 on Build GRCh38
          UniGene ID:
          Hs.420529
          Hs.744839
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s14600

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBE2V1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001032288.2 NP_001027459.1 3 351
          NM_001257393.1 NP_001244322.1 4 526
          NM_001257394.1 NP_001244323.1 2 585
          NM_001257395.1 NP_001244324.1 3 342
          NM_001257397.1 NP_001244326.1 4 434
          NM_001257398.1 NP_001244327.1 4 490
          NM_001257399.1 NP_001244328.1 4 469
          NM_001282575.2 NP_001269504.1 4 786
          NM_001282576.1 NP_001269505.1 4 625
          NM_001282577.2 NP_001269506.1 4 403
          NM_001282578.2 NP_001269507.1 3 540
          NM_001282579.1 NP_001269508.1 4 451
          NM_021988.5 NP_068823.2 3 734
          NM_022442.5 NP_071887.1 3 377
          NM_199144.2 NP_954595.1 4 657
          NR_047554.1 2 202
          GenBank mRNA AK314754.1 - 3 722
          AL110132.1 - 2 1152
          AY008273.1 - 3 350
          BC000468.2 - 3 291 2961017
          BC005215.1 - 4 784
          BC008944.2 - 2 268
          BT007382.1 - 3 281
          BU540046.1 - 4 658
          BU595183.1 - 328
          BX647511.1 - 2 170
          BX648147.1 - 2 176
          CA453411.1 - 375
          CD103154.1 - 2 514
          CN346855.1 - 4 458
          DA087027.1 - 4 351
          DA519011.1 - 3 351
          DA573591.1 - 4 421
          DA625165.1 - 4 436
          DA642098.1 - 4 567
          DA885410.1 - 3 294
          DA892136.1 - 4 428
          DA916492.1 - 3 492
          DB448941.1 - 4 403
          HY019085.1 - 4 423
          U39360.1 - 4 419
          U97280.1 - 3 294
          U97281.1 - 2 201
          Gene Symbol: TMEM189-UBE2V1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199203.2 NP_954673.1 7 1111
          GenBank mRNA BU156219.1 - 3 286
          BU619725.1 - 1 84
          U39361.1 - 4 572

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          regulation of DNA repair
          postreplication repair
          regulation of transcription, DNA-templated
          cell differentiation
          Fc-epsilon receptor signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription, DNA-templated
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          nucleotide-binding oligomerization domain containing signaling pathway
          protein K63-linked ubiquitination
          protein binding
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qlhjl:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0