Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR4A3 SIRNA Assays ›
          Gene Symbol:
          NR4A3
          Gene Name:
          nuclear receptor subfamily 4 group A member 3
          Gene Aliases:
          CHN, CSMF, MINOR, NOR1, TEC
          Chromosome Location:
          Chr.9: 99821855 - 99866893 on Build GRCh38
          UniGene ID:
          Hs.279522
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s15542
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR4A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006981.3 NP_008912.2 3 954
          NM_173199.2 NP_775291.1 3 954
          NM_173200.2 NP_775292.1 4 1026
          XM_017015162.1 XP_016870651.1 2 886
          GenBank mRNA AK226108.1 - 3 936
          D78579.1 - 3 956
          S81243.1 - 3 841
          U12767.1 - 2 317
          X89894.1 - 3 912

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00235001_m1

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          mesoderm formation
          transcription initiation from RNA polymerase II promoter
          gastrulation
          axon guidance
          biological_process
          pyruvate oxidation
          positive regulation of cardiac muscle hypertrophy
          positive regulation of glucose transport
          hippocampus development
          intracellular receptor signaling pathway
          adult behavior
          organ regeneration
          positive regulation of mast cell cytokine production
          common myeloid progenitor cell proliferation
          positive regulation of mast cell activation by Fc-epsilon receptor signaling pathway
          response to hydrogen peroxide
          mast cell degranulation
          steroid hormone mediated signaling pathway
          negative regulation of neuron apoptotic process
          cellular response to leptin stimulus
          cellular respiration
          fat cell differentiation
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          regulation of smooth muscle cell proliferation
          positive regulation of smooth muscle cell proliferation
          semicircular canal morphogenesis
          positive regulation of epithelial cell proliferation
          neuromuscular process controlling balance
          vestibular reflex
          regulation of type B pancreatic cell proliferation
          cellular response to corticotropin-releasing hormone stimulus
          cellular response to catecholamine stimulus
          energy homeostasis
          positive regulation of monocyte aggregation
          negative regulation of hydrogen peroxide-induced neuron death
          positive regulation of leukocyte apoptotic process
          positive regulation of feeding behavior
          regulation of energy homeostasis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription coactivator binding
          DNA binding
          steroid hormone receptor activity
          thyroid hormone receptor activity
          protein binding
          zinc ion binding
          protein kinase binding
          histone acetyltransferase binding
          glucocorticoid receptor binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5mtvh:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0