Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PICALM SIRNA Assays ›
Gene Symbol:
PICALM
Gene Name:
phosphatidylinositol binding clathrin assembly protein
Gene Aliases:
CALM, CLTH, LAP
Chromosome Location:
Chr.11: 85957171 - 86069881 on Build GRCh38
UniGene ID:
Hs.163893
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15799
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PICALM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008660.2
NP_001008660.1
3, 4
674
NM_001206946.1
NP_001193875.1
3, 4
674
NM_001206947.1
NP_001193876.1
3, 4
389
NM_007166.3
NP_009097.2
3, 4
674
XM_005274322.2
XP_005274379.1
3, 4
658
XM_005274323.2
XP_005274380.1
3, 4
658
XM_005274324.2
XP_005274381.1
3, 4
658
XM_005274325.2
XP_005274382.1
3, 4
658
XM_005274326.2
XP_005274383.1
3, 4
658
XM_005274327.2
XP_005274384.1
3, 4
658
XM_005274328.2
XP_005274385.1
3, 4
658
XM_005274329.3
XP_005274386.1
3, 4
658
XM_005274330.2
XP_005274387.1
3, 4
658
XM_005274331.2
XP_005274388.1
3, 4
658
XM_005274332.2
XP_005274389.1
3, 4
658
XM_005274333.2
XP_005274390.1
3, 4
658
XM_005274334.2
XP_005274391.1
3, 4
658
XM_005274335.2
XP_005274392.1
3, 4
658
XM_005274336.2
XP_005274393.1
3, 4
658
XM_005274337.2
XP_005274394.1
3, 4
658
XM_005274340.2
XP_005274397.1
3, 4
658
XM_006718699.2
XP_006718762.1
3, 4
658
XM_006718700.2
XP_006718763.1
3, 4
658
XM_006718701.3
XP_006718764.1
3, 4
658
XM_011545293.1
XP_011543595.1
3, 4
658
XM_017018381.1
XP_016873870.1
3, 4
658
XM_017018382.1
XP_016873871.1
3, 4
658
XM_017018383.1
XP_016873872.1
3, 4
658
XM_017018384.1
XP_016873873.1
3, 4
658
XM_017018385.1
XP_016873874.1
3, 4
658
XM_017018386.1
XP_016873875.1
3, 4
658
XM_017018387.1
XP_016873876.1
3, 4
658
XM_017018388.1
XP_016873877.1
3, 4
658
GenBank mRNA
AB210017.1
-
3, 4
674
AB451308.1
-
3, 4
357
AK291414.1
-
4, 5
541
AK300275.1
-
3, 4
366
BC048259.1
-
3, 4
435
5537605
BC064357.1
-
3, 4
636
5112878
BC073961.1
-
3, 4
510
6183858
DC300074.1
-
3, 4
389
U45976.1
-
3, 4
504
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
receptor-mediated endocytosis
cell proliferation
negative regulation of gene expression
synaptic vesicle maturation
vesicle-mediated transport
endosomal transport
regulation of endocytosis
receptor internalization
regulation of protein localization
cargo loading into vesicle
positive regulation of transcription, DNA-templated
negative regulation of receptor-mediated endocytosis
clathrin coat assembly
iron ion homeostasis
clathrin-mediated endocytosis
iron ion import into cell
cell-cell adhesion
positive regulation of neuron death
positive regulation of beta-amyloid formation
regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
protein binding
1-phosphatidylinositol binding
clathrin binding
clathrin heavy chain binding
clathrin adaptor activity
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0