Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1G SIRNA Assays ›
Gene Symbol:
CACNA1G
Gene Name:
calcium voltage-gated channel subunit alpha1 G
Gene Aliases:
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location:
Chr.17: 50560929 - 50629625 on Build GRCh38
UniGene ID:
Hs.591169
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17052
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256324.1
NP_001243253.1
9
2469
NM_001256325.1
NP_001243254.1
9
2469
NM_001256326.1
NP_001243255.1
9
2469
NM_001256327.1
NP_001243256.1
9
2469
NM_001256328.1
NP_001243257.1
9
2469
NM_001256329.1
NP_001243258.1
9
2469
NM_001256330.1
NP_001243259.1
9
2469
NM_001256331.1
NP_001243260.1
9
2469
NM_001256332.1
NP_001243261.1
9
2469
NM_001256333.1
NP_001243262.1
9
2469
NM_001256334.1
NP_001243263.1
9
2469
NM_001256359.1
NP_001243288.1
9
2469
NM_001256360.1
NP_001243289.1
9
2469
NM_001256361.1
NP_001243290.1
9
2469
NM_018896.4
NP_061496.2
9
2469
NM_198376.2
NP_938190.1
9
2469
NM_198377.2
NP_938191.2
9
2469
NM_198378.2
NP_938192.1
9
2469
NM_198379.2
NP_938193.1
9
2469
NM_198380.2
NP_938194.1
9
2469
NM_198382.2
NP_938196.1
9
2469
NM_198383.2
NP_938197.1
9
2469
NM_198384.2
NP_938198.1
9
2469
NM_198385.2
NP_938199.1
9
2469
NM_198386.2
NP_938200.1
9
2469
NM_198387.2
NP_938201.1
9
2469
NM_198388.2
NP_938202.1
9
2469
NM_198396.2
NP_938406.1
9
2469
NR_046054.1
9
2469
NR_046055.1
9
2469
NR_046056.1
9
2469
NR_046057.1
9
2469
NR_046058.1
9
2469
XM_006722160.3
XP_006722223.1
9
2608
XM_006722161.3
XP_006722224.1
9
2557
GenBank mRNA
AB012043.1
-
9
2105
AB032949.2
-
2
431
AF124351.1
-
9
2449
AF126965.1
-
9
2252
AF126966.1
-
9
2252
AF134985.1
-
9
2077
AF134986.1
-
9
2077
AF190860.1
-
9
2293
AF227744.1
-
9
2077
AF227745.1
-
9
2077
AF227746.1
-
9
2077
AF227747.1
-
9
2077
AF227748.1
-
9
2077
AF227749.1
-
9
2077
AF227750.1
-
9
2077
AF227751.1
-
9
2077
BC110995.1
-
9
2469
5502027
DQ494449.1
-
9
2077
DQ494450.1
-
9
2077
DQ494451.1
-
9
2077
DQ494452.1
-
9
2077
DQ494453.1
-
9
2077
DQ494454.1
-
9
2077
DQ494455.1
-
9
2077
DQ494456.1
-
9
2077
DQ494457.1
-
9
2077
DQ494458.1
-
9
2077
DQ494459.1
-
9
2077
DQ494460.1
-
9
2077
DQ494461.1
-
9
2077
DQ494462.1
-
9
2077
DQ494463.1
-
9
2077
DQ494464.1
-
9
2077
DQ494465.1
-
9
2077
DQ494466.1
-
9
2077
DQ494467.1
-
9
2077
DQ494468.1
-
9
2077
DQ494469.1
-
9
2077
DQ494470.1
-
9
2077
DQ494471.1
-
9
2077
DQ494472.1
-
9
2077
DQ494473.1
-
9
2077
DQ494474.1
-
9
2077
DQ494475.1
-
9
2077
DQ494476.1
-
9
2077
DQ494477.1
-
9
2077
DQ494478.1
-
9
2077
DQ494479.1
-
9
2077
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
response to nickel cation
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
SA node cell action potential
AV node cell action potential
SA node cell to atrial cardiac muscle cell signalling
AV node cell to bundle of His cell signaling
membrane depolarization during AV node cell action potential
membrane depolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
low voltage-gated calcium channel activity
voltage-gated calcium channel activity involved in AV node cell action potential
voltage-gated calcium channel activity involved SA node cell action potential
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0